The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001447.2 | NP_001438.1 | 9 - 10 | 8861 | 103 |
XM_006714761.3 | XP_006714824.1 | 10 - 11 | 9432 | 103 | |
XM_011537600.2 | XP_011535902.1 | 12 - 13 | 9200 | 103 | |
XM_011537603.2 | XP_011535905.1 | 10 - 11 | 9133 | 103 | |
XM_017009224.1 | XP_016864713.1 | 12 - 13 | 9202 | 103 | |
XM_017009225.1 | XP_016864714.1 | 11 - 12 | 9044 | 103 | |
XM_017009226.1 | XP_016864715.1 | 10 - 11 | 9437 | 103 | |
XM_017009227.1 | XP_016864716.1 | 10 - 11 | 9436 | 103 | |
GenBank mRNA | AF231022.1 | - | 9 - 10 | 8861 | 103 |