Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANK3
Entrez Gene ID
288
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases
ANKYRIN-G, MRT37
Chromosome Location
Chr.10: 60026298 - 60733526 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001149.3 NP_001140.2 8 - 9 1796 69
NM_001204403.1 NP_001191332.1 31 - 32 3934 69
NM_001204404.1 NP_001191333.1 31 - 32 4177 69
NM_001320874.1 NP_001307803.1 30 - 31 4191 69
NM_020987.4 NP_066267.2 30 - 31 4191 69
XM_005269715.3 XP_005269772.1 31 - 32 4078 69
XM_006717796.3 XP_006717859.1 32 - 33 4090 69
XM_006717802.3 XP_006717865.1 32 - 33 4090 69
XM_011539708.2 XP_011538010.1 32 - 33 4090 69
XM_011539709.2 XP_011538011.1 32 - 33 4090 69
XM_017016110.1 XP_016871599.1 32 - 33 4090 69
XM_017016111.1 XP_016871600.1 31 - 32 4078 69
XM_017016112.1 XP_016871601.1 32 - 33 4075 69
XM_017016113.1 XP_016871602.1 32 - 33 4090 69
XM_017016114.1 XP_016871603.1 32 - 33 4189 69
XM_017016115.1 XP_016871604.1 32 - 33 3919 69
XM_017016116.1 XP_016871605.1 32 - 33 4090 69
XM_017016117.1 XP_016871606.1 32 - 33 4090 69
XM_017016118.1 XP_016871607.1 32 - 33 4090 69
XM_017016119.1 XP_016871608.1 32 - 33 4090 69
XM_017016120.1 XP_016871609.1 32 - 33 4090 69
XM_017016121.1 XP_016871610.1 32 - 33 4090 69
XM_017016122.1 XP_016871611.1 30 - 31 4024 69
XM_017016123.1 XP_016871612.1 32 - 33 4090 69
XM_017016124.1 XP_016871613.1 32 - 33 4090 69
XM_017016125.1 XP_016871614.1 31 - 32 4063 69
XM_017016126.1 XP_016871615.1 30 - 31 3958 69
XM_017016127.1 XP_016871616.1 30 - 31 3958 69
XM_017016128.1 XP_016871617.1 32 - 33 4090 69
XM_017016129.1 XP_016871618.1 32 - 33 4090 69
XM_017016130.1 XP_016871619.1 30 - 31 4024 69
XM_017016131.1 XP_016871620.1 30 - 31 4009 69
XM_017016132.1 XP_016871621.1 30 - 31 4024 69
XM_017016133.1 XP_016871622.1 30 - 31 4123 69
XM_017016134.1 XP_016871623.1 30 - 31 3958 69
XM_017016135.1 XP_016871624.1 30 - 31 4123 69
XM_017016136.1 XP_016871625.1 32 - 33 4090 69
XM_017016137.1 XP_016871626.1 32 - 33 4090 69
XM_017016138.1 XP_016871627.1 32 - 33 4090 69
XM_017016139.1 XP_016871628.1 30 - 31 4024 69
XM_017016140.1 XP_016871629.1 30 - 31 4024 69
XM_017016141.1 XP_016871630.1 30 - 31 3958 69
XM_017016142.1 XP_016871631.1 29 - 30 3946 69
XM_017016143.1 XP_016871632.1 30 - 31 3958 69
XM_017016144.1 XP_016871633.1 30 - 31 3958 69
XM_017016145.1 XP_016871634.1 29 - 30 3946 69
XM_017016146.1 XP_016871635.1 29 - 30 3946 69
XM_017016147.1 XP_016871636.1 29 - 30 3946 69
XM_017016148.1 XP_016871637.1 30 - 31 3840 69
XM_017016149.1 XP_016871638.1 29 - 30 4045 69
XM_017016150.1 XP_016871639.1 30 - 31 4024 69
GenBank mRNA AB209308.1 - 31 - 32 4079 69
AK092527.1 - 8 - 9 1801 69
AK126851.1 - 12 - 13 1879 69
AK292927.1 - 8 - 9 1682 69
AK295661.1 - 31 - 32 3884 69
AL136710.1 - 8 - 9 1455 69
BX537917.1 - 31 - 32 3805 69
BX648574.1 - 31 - 32 3899 69
U13616.1 - 30 - 31 3937 69

View Full Product Details