Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF10
Entrez Gene ID
9921
Gene Name
ring finger protein 10
Gene Aliases
RIE2
Chromosome Location
Chr.12: 120534309 - 120577594 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014868.4 NP_055683.3 10 - 11 2147 70
XM_005254013.2 XP_005254070.1 11 - 12 2239 70
XM_006719717.2 XP_006719780.1 10 - 11 2182 70
XM_017020282.1 XP_016875771.1 11 - 12 2224 70
XM_017020283.1 XP_016875772.1 10 - 11 2182 70
GenBank mRNA AB027196.1 - 10 - 11 2111 70
AK094011.1 - 11 - 12 2048 70
AK097807.1 - 11 - 12 2073 70
AK126274.1 - 11 - 12 1952 70
AK301608.1 - 11 - 12 2021 70
AK315558.1 - 10 - 11 2129 70
AL389976.1 - 9 - 10 1419 70
BC016622.1 - 10 - 11 2111 70
BC031596.1 - 10 - 11 2111 70
BC101709.1 - 10 - 11 1737 70
BC101715.1 - 10 - 11 1737 70
BC143493.1 - 10 - 11 1736 70
BT006974.1 - 10 - 11 1664 70
D87451.2 - 10 - 11 1664 70

View Full Product Details