The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014868.4 | NP_055683.3 | 16 - 17 | 2841 | 88 |
XM_005254013.2 | XP_005254070.1 | 17 - 18 | 2933 | 88 | |
XM_006719717.2 | XP_006719780.1 | 16 - 17 | 2876 | 88 | |
XM_017020282.1 | XP_016875771.1 | 17 - 18 | 2918 | 88 | |
GenBank mRNA | AB027196.1 | - | 16 - 17 | 2805 | 88 |
AK094011.1 | - | 17 - 18 | 2742 | 88 | |
AK097807.1 | - | 17 - 18 | 2767 | 88 | |
AK126274.1 | - | 17 - 18 | 2646 | 88 | |
AK301608.1 | - | 17 - 18 | 2715 | 88 | |
AK315558.1 | - | 16 - 17 | 2823 | 88 | |
AL389976.1 | - | 15 - 16 | 2113 | 88 | |
BC016622.1 | - | 16 - 17 | 2805 | 88 | |
BC031596.1 | - | 16 - 17 | 2805 | 88 | |
BC101709.1 | - | 16 - 17 | 2431 | 88 | |
BC101715.1 | - | 16 - 17 | 2431 | 88 | |
BC143493.1 | - | 15 - 16 | 2329 | 88 | |
BT006974.1 | - | 16 - 17 | 2358 | 88 | |
D87451.2 | - | 16 - 17 | 2358 | 88 |