The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144625.4 | NP_653226.4 | 25 - 26 | 3259 | 75 |
XM_006711736.3 | XP_006711799.1 | 26 - 27 | 3289 | 75 | |
XM_011544085.2 | XP_011542387.1 | 23 - 24 | 2688 | 75 | |
XM_011544086.2 | XP_011542388.1 | 24 - 25 | 2690 | 75 | |
XM_017000315.1 | XP_016855804.1 | 26 - 27 | 3024 | 75 | |
GenBank mRNA | AK057540.1 | - | 13 - 14 | 1390 | 75 |
AK122927.1 | - | 25 - 26 | 3099 | 75 | |
AY327404.1 | - | 15 - 16 | 1864 | 75 |