The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144625.4 | NP_653226.4 | 10 - 11 | 1497 | 132 |
XM_006711736.3 | XP_006711799.1 | 11 - 12 | 1527 | 132 | |
XM_011544085.2 | XP_011542387.1 | 8 - 9 | 926 | 132 | |
XM_011544086.2 | XP_011542388.1 | 9 - 10 | 928 | 132 | |
XM_011544087.2 | XP_011542389.1 | 11 - 12 | 1527 | 132 | |
XM_011544091.1 | XP_011542393.1 | 11 - 12 | 1527 | 132 | |
XM_011544092.2 | XP_011542394.1 | 11 - 12 | 1527 | 132 | |
XM_017000315.1 | XP_016855804.1 | 11 - 12 | 1262 | 132 | |
GenBank mRNA | AK122927.1 | - | 10 - 11 | 1495 | 132 |
AY327404.1 | - | 5 - 6 | 603 | 132 |