The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174152.1 | NP_001167623.1 | 2 - 3 | 162 | 79 |
NM_001174153.1 | NP_001167624.1 | 2 - 3 | 162 | 79 | |
NM_005833.3 | NP_005824.2 | 1 - 2 | 323 | 79 | |
XM_017014177.1 | XP_016869666.1 | 2 - 3 | 147 | 79 | |
XM_017014178.1 | XP_016869667.1 | 2 - 3 | 211 | 79 | |
XM_017014180.1 | XP_016869669.1 | 2 - 3 | 211 | 79 | |
XM_017014181.1 | XP_016869670.1 | 1 - 2 | 322 | 79 | |
XM_017014182.1 | XP_016869671.1 | 2 - 3 | 147 | 79 | |
XM_017014183.1 | XP_016869672.1 | 1 - 2 | 322 | 79 | |
GenBank mRNA | AK290768.1 | - | 1 - 2 | 167 | 79 |
AL832249.1 | - | 2 - 3 | 159 | 79 | |
BC000503.2 | - | 1 - 2 | 300 | 79 | |
BC047023.1 | - | 2 - 3 | 142 | 79 | |
BC053541.1 | - | 1 - 2 | 132 | 79 | |
BC065725.1 | - | 2 - 3 | 165 | 79 | |
CX758286.1 | - | 1 - 2 | 323 | 79 | |
Z97074.1 | - | 2 - 3 | 142 | 79 |