The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198274.3 | NP_938015.1 | 4 - 5 | 748 | 125 |
GenBank mRNA | AK091724.1 | - | 4 - 5 | 666 | 125 |
AL832035.1 | - | 4 - 5 | 665 | 125 | |
AY518933.1 | - | 4 - 5 | 645 | 125 | |
BC126191.1 | - | 4 - 5 | 748 | 125 | |
BX647878.1 | - | 4 - 5 | 667 | 125 |