Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA3
Entrez Gene ID
1136
Gene Name
cholinergic receptor nicotinic alpha 3 subunit
Gene Aliases
LNCR2, NACHRA3, PAOD2
Chromosome Location
Chr.15: 78593052 - 78621295 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166694.1 NP_001160166.1 5 - 6 1883 95
GenBank mRNA BC006114.1 - 5 - 6 1883 95
BT006646.1 - 5 - 6 1382 95

View Full Product Details