The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019852.4 | NP_062826.2 | 3 - 4 | 877 | 87 |
XM_011536968.1 | XP_011535270.1 | 3 - 4 | 872 | 87 | |
GenBank mRNA | AK000222.1 | - | 3 - 4 | 800 | 87 |
AK093509.1 | - | 1 - 2 | 659 | 87 | |
AK098655.1 | - | 2 - 3 | 573 | 87 | |
AK304251.1 | - | 3 - 4 | 842 | 87 | |
AK304570.1 | - | 4 - 5 | 628 | 87 | |
BC001650.2 | - | 3 - 4 | 811 | 87 | |
BC003031.1 | - | 3 - 4 | 811 | 87 | |
BC052244.1 | - | 3 - 4 | 761 | 87 | |
BX247964.1 | - | 3 - 4 | 318 | 87 |