The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152435.2 | NP_689648.2 | 3 - 4 | 422 | 75 |
GenBank mRNA | AB075878.1 | - | 3 - 4 | 377 | 75 |
AK054617.1 | - | 3 - 4 | 422 | 75 | |
AK122802.1 | - | 3 - 4 | 389 | 75 | |
AK290828.1 | - | 3 - 4 | 377 | 75 | |
BC029146.1 | - | 3 - 4 | 358 | 75 |