Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBC1D24
Entrez Gene ID
57465
Gene Name
TBC1 domain family member 24
Gene Aliases
DFNA65, DFNB86, DOORS, EIEE16, FIME, TLDC6
Chromosome Location
Chr.16: 2475104 - 2509669 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199107.1 NP_001186036.1 3 - 4 1130 73
XM_017023493.1 XP_016878982.1 3 - 4 1172 73
GenBank mRNA AB032997.1 - 3 - 4 1123 73
AB449911.1 - 2 - 3 990 73
BC127015.1 - 2 - 3 1044 73

View Full Product Details