The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006342.2 | NP_006333.1 | 11 - 12 | 2170 | 65 |
XM_005247929.2 | XP_005247986.1 | 11 - 12 | 2180 | 65 | |
XM_005247930.2 | XP_005247987.1 | 10 - 11 | 1209 | 65 | |
XM_011513386.2 | XP_011511688.1 | 11 - 12 | 2180 | 65 | |
XM_017007653.1 | XP_016863142.1 | 11 - 12 | 2222 | 65 | |
GenBank mRNA | AF093543.1 | - | 11 - 12 | 2123 | 65 |
AF479620.1 | - | 10 - 11 | 1036 | 65 | |
AJ243997.1 | - | 11 - 12 | 2115 | 65 | |
BC106071.1 | - | 11 - 12 | 2131 | 65 | |
BC111771.1 | - | 11 - 12 | 2083 | 65 |