Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACC3
Entrez Gene ID
10460
Gene Name
transforming acidic coiled-coil containing protein 3
Gene Aliases
ERIC-1, ERIC1
Chromosome Location
Chr.4: 1721383 - 1745178 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006342.2 NP_006333.1 12 - 13 2217 65
XM_005247929.2 XP_005247986.1 12 - 13 2227 65
XM_005247930.2 XP_005247987.1 11 - 12 1256 65
XM_017007653.1 XP_016863142.1 12 - 13 2269 65
GenBank mRNA AF093543.1 - 12 - 13 2170 65
AF479620.1 - 11 - 12 1083 65
AJ243997.1 - 12 - 13 2162 65
BC106071.1 - 12 - 13 2178 65
BC111771.1 - 12 - 13 2130 65

View Full Product Details