Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACC3
Entrez Gene ID
10460
Gene Name
transforming acidic coiled-coil containing protein 3
Gene Aliases
ERIC-1, ERIC1
Chromosome Location
Chr.4: 1721383 - 1745178 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006342.2 NP_006333.1 1 - 2 151 119
XM_005247929.2 XP_005247986.1 1 - 2 149 119
XM_005247930.2 XP_005247987.1 1 - 2 258 119
XM_011513386.2 XP_011511688.1 1 - 2 149 119
GenBank mRNA AF093543.1 - 1 - 2 104 119
AF479620.1 - 1 - 2 96 119
AJ243997.1 - 1 - 2 96 119
AK225631.1 - 1 - 2 119 119
AK302463.1 - 1 - 2 122 119
BC106071.1 - 1 - 2 112 119
BC111771.1 - 1 - 2 64 119
DB098941.1 - 1 - 2 149 119

View Full Product Details