The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004706.3 | NP_004697.2 | 17 - 18 | 1731 | 106 |
NM_198977.1 | NP_945328.1 | 16 - 17 | 1632 | 106 | |
NM_199002.1 | NP_945353.1 | 17 - 18 | 1756 | 106 | |
XM_005259386.4 | XP_005259443.1 | 19 - 20 | 2263 | 106 | |
XM_005259390.4 | XP_005259447.1 | 17 - 18 | 2079 | 106 | |
XM_011527468.2 | XP_011525770.1 | 18 - 19 | 2091 | 106 | |
GenBank mRNA | AL832294.1 | - | 8 - 9 | 895 | 106 |
BC005155.2 | - | 14 - 15 | 1469 | 106 | |
BC011726.2 | - | 16 - 17 | 1607 | 106 | |
BC034013.1 | - | 17 - 18 | 1687 | 106 | |
BC035127.1 | - | 4 - 5 | 1269 | 106 | |
BC067262.1 | - | 16 - 17 | 1575 | 106 | |
BT007421.1 | - | 14 - 15 | 1445 | 106 | |
U64105.1 | - | 17 - 18 | 1676 | 106 |