The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016297.3 | NP_057381.3 | 1 - 2 | 180 | 57 |
GenBank mRNA | AB020715.1 | - | 1 - 2 | 118 | 57 |
AF181490.1 | - | 1 - 2 | 169 | 57 | |
AK225302.1 | - | 1 - 2 | 130 | 57 | |
AK303043.1 | - | 1 - 2 | 130 | 57 | |
AK314453.1 | - | 1 - 2 | 132 | 57 | |
AY359063.1 | - | 1 - 2 | 133 | 57 | |
BC022259.1 | - | 1 - 2 | 130 | 57 | |
BC033815.1 | - | 1 - 2 | 133 | 57 | |
BC051891.1 | - | 1 - 2 | 167 | 57 | |
DC417137.1 | - | 1 - 2 | 180 | 57 |