Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GJA8
Entrez Gene ID
2703
Gene Name
gap junction protein alpha 8
Gene Aliases
CAE, CAE1, CTRCT1, CX50, CZP1, MP70
Chromosome Location
Chr.1: 147902820 - 147915287 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005267.4 NP_005258.2 1 - 2 41 63
GenBank mRNA CD674975.1 - 1 - 2 41 63

View Full Product Details