The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152384.2 | NP_689597.1 | 11 - 12 | 986 | 74 |
GenBank mRNA | AK097935.1 | - | 3 - 4 | 699 | 74 |
AK312680.1 | - | 10 - 11 | 903 | 74 | |
AL834305.1 | - | 11 - 12 | 977 | 74 | |
AY604003.1 | - | 11 - 12 | 1002 | 74 | |
AY604004.1 | - | 10 - 11 | 939 | 74 | |
BC044593.1 | - | 11 - 12 | 983 | 74 | |
H09195.1 | - | 2 - 3 | 78 | 74 | |
KU178857.1 | - | 11 - 12 | 928 | 74 | |
KU178858.1 | - | 10 - 11 | 865 | 74 |