The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022140.3 | NP_071423.3 | 18 - 19 | 1899 | 99 |
XM_011543530.1 | XP_011541832.1 | 18 - 19 | 2076 | 99 | |
XM_011543531.2 | XP_011541833.1 | 18 - 19 | 2076 | 99 | |
XM_011543532.1 | XP_011541834.1 | 18 - 19 | 2076 | 99 | |
XM_011543533.1 | XP_011541835.1 | 17 - 18 | 1992 | 99 | |
XM_017009689.1 | XP_016865178.1 | 17 - 18 | 1992 | 99 | |
GenBank mRNA | AB030240.1 | - | 18 - 19 | 1631 | 99 |
AK096057.1 | - | 7 - 8 | 581 | 99 | |
BC114632.1 | - | 18 - 19 | 1725 | 99 | |
CB048933.1 | - | 5 - 6 | 454 | 99 |