The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022140.3 | NP_071423.3 | 3 - 4 | 532 | 157 |
XM_011543530.1 | XP_011541832.1 | 3 - 4 | 709 | 157 | |
XM_011543531.2 | XP_011541833.1 | 3 - 4 | 709 | 157 | |
XM_011543532.1 | XP_011541834.1 | 3 - 4 | 709 | 157 | |
XM_011543533.1 | XP_011541835.1 | 3 - 4 | 709 | 157 | |
XM_017009689.1 | XP_016865178.1 | 3 - 4 | 709 | 157 | |
GenBank mRNA | AB030240.1 | - | 3 - 4 | 261 | 157 |
BC031042.1 | - | 3 - 4 | 532 | 157 | |
BC114632.1 | - | 3 - 4 | 358 | 157 | |
BC114942.1 | - | 3 - 4 | 353 | 157 |