The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022140.3 | NP_071423.3 | 9 - 10 | 1073 | 98 |
XM_011543530.1 | XP_011541832.1 | 9 - 10 | 1250 | 98 | |
XM_011543531.2 | XP_011541833.1 | 9 - 10 | 1250 | 98 | |
XM_011543532.1 | XP_011541834.1 | 9 - 10 | 1250 | 98 | |
XM_011543533.1 | XP_011541835.1 | 9 - 10 | 1250 | 98 | |
XM_017009689.1 | XP_016865178.1 | 9 - 10 | 1250 | 98 | |
GenBank mRNA | AB030240.1 | - | 9 - 10 | 802 | 98 |
AK123285.1 | - | 5 - 6 | 1800 | 98 | |
BC114632.1 | - | 9 - 10 | 899 | 98 | |
BC114942.1 | - | 9 - 10 | 894 | 98 |