The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134477.2 | NP_001127949.1 | 21 - 22 | 1546 | 89 |
NM_001242992.1 | NP_001229921.1 | 20 - 21 | 1491 | 89 | |
NM_002582.3 | NP_002573.1 | 21 - 22 | 1629 | 89 | |
XM_011522510.2 | XP_011520812.1 | 21 - 22 | 1635 | 89 | |
XM_011522511.2 | XP_011520813.1 | 21 - 22 | 1634 | 89 | |
XM_011522513.2 | XP_011520815.1 | 21 - 22 | 1545 | 89 | |
XM_017023258.1 | XP_016878747.1 | 20 - 21 | 1555 | 89 | |
XM_017023259.1 | XP_016878748.1 | 17 - 18 | 1234 | 89 | |
XM_017023260.1 | XP_016878749.1 | 15 - 16 | 1123 | 89 | |
GenBank mRNA | AJ005698.1 | - | 21 - 22 | 1540 | 89 |
AK293189.1 | - | 21 - 22 | 1538 | 89 | |
AK299653.1 | - | 14 - 15 | 1071 | 89 | |
AK301648.1 | - | 20 - 21 | 1481 | 89 | |
AK315020.1 | - | 21 - 22 | 1625 | 89 | |
BC044763.1 | - | 21 - 22 | 1567 | 89 | |
BC050029.1 | - | 21 - 22 | 1567 | 89 |