Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARN
Entrez Gene ID
5073
Gene Name
poly(A)-specific ribonuclease
Gene Aliases
DAN, DKCB6, PFBMFT4
Chromosome Location
Chr.16: 14435700 - 14630277 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134477.2 NP_001127949.1 8 - 9 686 161
NM_001242992.1 NP_001229921.1 7 - 8 631 161
NM_002582.3 NP_002573.1 8 - 9 769 161
XM_011522510.2 XP_011520812.1 8 - 9 775 161
XM_011522511.2 XP_011520813.1 8 - 9 774 161
XM_011522513.2 XP_011520815.1 8 - 9 685 161
XM_011522514.1 XP_011520816.1 8 - 9 773 161
XM_017023258.1 XP_016878747.1 8 - 9 773 161
XM_017023259.1 XP_016878748.1 5 - 6 418 161
XM_017023260.1 XP_016878749.1 3 - 4 307 161
GenBank mRNA AJ005698.1 - 8 - 9 680 161
AK293189.1 - 8 - 9 678 161
AK301648.1 - 7 - 8 621 161
AK315020.1 - 8 - 9 765 161
BC044763.1 - 8 - 9 707 161
BC050029.1 - 8 - 9 707 161

View Full Product Details