The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172574.1 | NP_001166045.1 | 6 - 7 | 655 | 150 |
NM_001322042.1 | NP_001308971.1 | 6 - 7 | 655 | 150 | |
NM_001322043.1 | NP_001308972.1 | 6 - 7 | 649 | 150 | |
NM_001322045.1 | NP_001308974.1 | 6 - 7 | 715 | 150 | |
NM_024596.4 | NP_078872.2 | 6 - 7 | 655 | 150 | |
NR_136159.1 | 5 - 6 | 540 | 150 | ||
XM_011534755.2 | XP_011533057.1 | 6 - 7 | 655 | 150 | |
XM_011534756.2 | XP_011533058.1 | 6 - 7 | 654 | 150 | |
XM_011534757.2 | XP_011533059.1 | 6 - 7 | 654 | 150 | |
XM_011534758.2 | XP_011533060.1 | 6 - 7 | 654 | 150 | |
XM_011534759.2 | XP_011533061.1 | 6 - 7 | 654 | 150 | |
XM_011534760.2 | XP_011533062.1 | 2 - 3 | 231 | 150 | |
XM_017013829.1 | XP_016869318.1 | 6 - 7 | 655 | 150 | |
XM_017013830.1 | XP_016869319.1 | 6 - 7 | 654 | 150 | |
XM_017013831.1 | XP_016869320.1 | 6 - 7 | 654 | 150 | |
XM_017013832.1 | XP_016869321.1 | 6 - 7 | 654 | 150 | |
XM_017013833.1 | XP_016869322.1 | 6 - 7 | 654 | 150 | |
XM_017013834.1 | XP_016869323.1 | 6 - 7 | 654 | 150 | |
GenBank mRNA | AK022909.1 | - | 3 - 4 | 294 | 150 |
AK308387.1 | - | 6 - 7 | 620 | 150 | |
BC030702.1 | - | 6 - 7 | 652 | 150 |