Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRVI1
Entrez Gene ID
10335
Gene Name
murine retrovirus integration site 1 homolog
Gene Aliases
IRAG, JAW1L
Chromosome Location
Chr.11: 10573091 - 10693988 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206881.1 NP_001193810.1 6 - 7 919 85

View Full Product Details