The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206665.2 | NP_001193594.1 | 5 - 6 | 1389 | 79 |
NM_001320247.1 | NP_001307176.1 | 4 - 5 | 1531 | 79 | |
NM_052868.5 | NP_443100.1 | 4 - 5 | 1531 | 79 | |
XM_006711631.3 | XP_006711694.1 | 4 - 5 | 1280 | 79 | |
XM_011510159.2 | XP_011508461.1 | 4 - 5 | 1911 | 79 | |
XM_017002835.1 | XP_016858324.1 | 5 - 6 | 1395 | 79 | |
XM_017002836.1 | XP_016858325.1 | 5 - 6 | 1586 | 79 | |
XM_017002837.1 | XP_016858326.1 | 5 - 6 | 1395 | 79 | |
XM_017002838.1 | XP_016858327.1 | 4 - 5 | 3531 | 79 | |
XM_017002839.1 | XP_016858328.1 | 5 - 6 | 3443 | 79 | |
XM_017002840.1 | XP_016858329.1 | 5 - 6 | 1557 | 79 | |
GenBank mRNA | AF311906.1 | - | 4 - 5 | 1360 | 79 |
AF407274.1 | - | 4 - 5 | 1471 | 79 | |
AK055843.1 | - | 4 - 5 | 79 | ||
AK308115.1 | - | 5 - 6 | 1390 | 79 | |
AY044845.1 | - | 4 - 5 | 1386 | 79 | |
AY157579.1 | - | 4 - 5 | 1340 | 79 | |
BC004108.2 | - | 3 - 4 | 928 | 79 | |
BC011896.2 | - | 1 - 2 | 409 | 79 | |
BC023649.2 | - | 2 - 3 | 596 | 79 | |
BC053881.1 | - | 4 - 5 | 1477 | 79 |