The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002133.2 | NP_002124.1 | 3 - 4 | 742 | 82 |
GenBank mRNA | AK313120.1 | - | 3 - 4 | 712 | 82 |
BC001491.2 | - | 3 - 4 | 701 | 82 | |
BT019785.1 | - | 2 - 3 | 635 | 82 | |
CR456505.1 | - | 3 - 4 | 750 | 82 | |
CR541945.1 | - | 3 - 4 | 635 | 82 | |
CR541968.1 | - | 3 - 4 | 635 | 82 | |
JF323038.1 | - | 3 - 4 | 639 | 82 | |
X06985.1 | - | 3 - 4 | 715 | 82 |