The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145353.1 | NP_001138825.1 | 4 - 5 | 688 | 149 |
NM_172373.3 | NP_758961.1 | 5 - 6 | 905 | 149 | |
XM_005266276.1 | XP_005266333.1 | 5 - 6 | 794 | 149 | |
XM_005266277.3 | XP_005266334.1 | 5 - 6 | 1337 | 149 | |
XM_011534950.1 | XP_011533252.1 | 7 - 8 | 1143 | 149 | |
XM_011534951.2 | XP_011533253.1 | 6 - 7 | 1456 | 149 | |
XM_017020409.1 | XP_016875898.1 | 7 - 8 | 1123 | 149 | |
XM_017020410.1 | XP_016875899.1 | 6 - 7 | 1060 | 149 | |
XM_017020411.1 | XP_016875900.1 | 6 - 7 | 1006 | 149 | |
XM_017020412.1 | XP_016875901.1 | 6 - 7 | 1134 | 149 | |
XM_017020413.1 | XP_016875902.1 | 6 - 7 | 1188 | 149 | |
GenBank mRNA | AK304289.1 | - | 4 - 5 | 631 | 149 |
BC030507.2 | - | 4 - 5 | 760 | 149 | |
M82882.1 | - | 5 - 6 | 847 | 149 |