The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201427.1 | NP_001188356.1 | 22 - 23 | 2819 | 90 |
XM_006715039.3 | XP_006715102.3 | 23 - 24 | 3491 | 90 | |
XM_006715040.3 | XP_006715103.3 | 22 - 23 | 3463 | 90 | |
XM_006715042.2 | XP_006715105.1 | 23 - 24 | 5486 | 90 | |
XM_006715043.2 | XP_006715106.1 | 23 - 24 | 2863 | 90 | |
XM_006715045.3 | XP_006715108.1 | 23 - 24 | 4078 | 90 | |
XM_006715046.3 | XP_006715109.1 | 23 - 24 | 3256 | 90 | |
XM_017010630.1 | XP_016866119.1 | 23 - 24 | 4102 | 90 | |
GenBank mRNA | AB002379.2 | - | 22 - 23 | 2819 | 90 |
BX647839.1 | - | 6 - 7 | 3401 | 90 |