The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138961.2 | NP_620411.2 | 1 - 2 | 243 | 84 |
GenBank mRNA | AF361746.1 | - | 1 - 2 | 208 | 84 |
AK075396.1 | - | 1 - 2 | 206 | 84 | |
AK091023.1 | - | 1 - 2 | 1158 | 84 | |
AK314710.1 | - | 1 - 2 | 206 | 84 | |
AY358382.1 | - | 1 - 2 | 195 | 84 | |
BC016868.1 | - | 1 - 2 | 77 | 84 | |
DA257581.1 | - | 1 - 2 | 243 | 84 |