The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316350.1 | NP_001303279.1 | 5 - 6 | 794 | 108 |
NM_014236.3 | NP_055051.1 | 6 - 7 | 977 | 108 | |
XM_005273313.4 | XP_005273370.1 | 6 - 7 | 937 | 108 | |
XM_011544303.2 | XP_011542605.1 | 7 - 8 | 1026 | 108 | |
XM_011544304.2 | XP_011542606.1 | 6 - 7 | 845 | 108 | |
GenBank mRNA | AB209882.1 | - | 6 - 7 | 882 | 108 |
AF043937.1 | - | 6 - 7 | 853 | 108 | |
AJ002190.1 | - | 6 - 7 | 928 | 108 | |
AK222773.1 | - | 6 - 7 | 933 | 108 | |
AK223304.1 | - | 6 - 7 | 936 | 108 | |
AK297982.1 | - | 5 - 6 | 755 | 108 | |
AK311386.1 | - | 5 - 6 | 627 | 108 | |
AK314181.1 | - | 6 - 7 | 940 | 108 | |
BC000450.2 | - | 6 - 7 | 824 | 108 |