The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080656.2 | NP_542387.1 | 1 - 2 | 288 | 71 |
GenBank mRNA | BC008293.1 | - | 1 - 2 | 279 | 71 |