Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAZ1B
Entrez Gene ID
9031
Gene Name
bromodomain adjacent to zinc finger domain 1B
Gene Aliases
WBSCR10, WBSCR9, WSTF
Chromosome Location
Chr.7: 73440398 - 73522298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032408.3 NP_115784.1 10 - 11 3316 71
XM_017012773.1 XP_016868262.1 10 - 11 3329 71
GenBank mRNA AB032253.1 - 10 - 11 3309 71
AF072810.1 - 10 - 11 3316 71
AF084479.1 - 10 - 11 3248 71
BC136520.1 - 10 - 11 3149 71

View Full Product Details