Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EVI5
Entrez Gene ID
7813
Gene Name
ecotropic viral integration site 5
Gene Aliases
EVI-5, NB4S
Chromosome Location
Chr.1: 92508696 - 92792404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308248.1 NP_001295177.1 18 - 19 2166 112
NM_005665.5 NP_005656.4 17 - 18 2133 112
XM_017002272.1 XP_016857761.1 20 - 21 2392 112
XM_017002278.1 XP_016857767.1 19 - 20 2245 112
XM_017002280.1 XP_016857769.1 19 - 20 2535 112
XM_017002281.1 XP_016857770.1 18 - 19 2466 112
XM_017002284.1 XP_016857773.1 18 - 19 2387 112
GenBank mRNA AB449875.1 - 18 - 19 2156 112

View Full Product Details