The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308248.1 | NP_001295177.1 | 3 - 4 | 477 | 159 |
NM_005665.5 | NP_005656.4 | 3 - 4 | 477 | 159 | |
XM_017002271.1 | XP_016857760.1 | 3 - 4 | 475 | 159 | |
XM_017002276.1 | XP_016857765.1 | 3 - 4 | 699 | 159 | |
XM_017002277.1 | XP_016857766.1 | 3 - 4 | 475 | 159 | |
XM_017002280.1 | XP_016857769.1 | 3 - 4 | 699 | 159 | |
XM_017002281.1 | XP_016857770.1 | 3 - 4 | 777 | 159 | |
XM_017002283.1 | XP_016857772.1 | 3 - 4 | 475 | 159 | |
XM_017002284.1 | XP_016857773.1 | 3 - 4 | 698 | 159 | |
XM_017002285.1 | XP_016857774.1 | 3 - 4 | 698 | 159 | |
XM_017002286.1 | XP_016857775.1 | 2 - 3 | 469 | 159 | |
GenBank mRNA | AB449875.1 | - | 3 - 4 | 467 | 159 |
AF008915.1 | - | 3 - 4 | 478 | 159 |