Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EVI5
Entrez Gene ID
7813
Gene Name
ecotropic viral integration site 5
Gene Aliases
EVI-5, NB4S
Chromosome Location
Chr.1: 92508696 - 92792404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308248.1 NP_001295177.1 8 - 9 1141 82
NM_005665.5 NP_005656.4 8 - 9 1141 82
XM_017002269.1 XP_016857758.1 9 - 10 1220 82
XM_017002270.1 XP_016857759.1 9 - 10 1531 82
XM_017002271.1 XP_016857760.1 8 - 9 1139 82
XM_017002272.1 XP_016857761.1 9 - 10 1220 82
XM_017002273.1 XP_016857762.1 9 - 10 1446 82
XM_017002274.1 XP_016857763.1 9 - 10 1195 82
XM_017002275.1 XP_016857764.1 9 - 10 1183 82
XM_017002276.1 XP_016857765.1 8 - 9 1363 82
XM_017002277.1 XP_016857766.1 8 - 9 1139 82
XM_017002278.1 XP_016857767.1 9 - 10 1220 82
XM_017002279.1 XP_016857768.1 8 - 9 1132 82
XM_017002280.1 XP_016857769.1 8 - 9 1363 82
XM_017002281.1 XP_016857770.1 8 - 9 1441 82
XM_017002282.1 XP_016857771.1 9 - 10 1220 82
XM_017002283.1 XP_016857772.1 8 - 9 1139 82
XM_017002284.1 XP_016857773.1 8 - 9 1362 82
XM_017002285.1 XP_016857774.1 8 - 9 1362 82
XM_017002286.1 XP_016857775.1 7 - 8 1133 82
XM_017002287.1 XP_016857776.1 8 - 9 1214 82
XM_017002288.1 XP_016857777.1 6 - 7 756 82
GenBank mRNA AB209513.1 - 1 - 2 50 82
AB449875.1 - 8 - 9 1131 82
AF008915.1 - 8 - 9 1142 82

View Full Product Details