Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC41A3
Entrez Gene ID
54946
Gene Name
solute carrier family 41 member 3
Gene Aliases
SLC41A1-L2
Chromosome Location
Chr.3: 126006357 - 126101550 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK021925.1 - 6 - 7 711 62

View Full Product Details