The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001512.3 | NP_001503.1 | 6 - 7 | 690 | 147 |
XM_005249035.3 | XP_005249092.1 | 6 - 7 | 783 | 147 | |
XM_011514534.2 | XP_011512836.1 | 5 - 6 | 633 | 147 | |
XM_011514535.2 | XP_011512837.1 | 5 - 6 | 729 | 147 | |
GenBank mRNA | AF020918.1 | - | 6 - 7 | 610 | 147 |
AF025887.1 | - | 6 - 7 | 613 | 147 | |
AF125271.1 | - | 5 - 6 | 540 | 147 | |
AF125272.1 | - | 5 - 6 | 540 | 147 | |
AF125273.1 | - | 5 - 6 | 540 | 147 | |
AK054804.1 | - | 4 - 5 | 1047 | 147 | |
AK315369.1 | - | 6 - 7 | 589 | 147 | |
BC015523.2 | - | 5 - 6 | 560 | 147 | |
BC063439.1 | - | 4 - 5 | 527 | 147 | |
BG716171.1 | - | 6 - 7 | 659 | 146 | |
Y13047.1 | - | 5 - 6 | 540 | 147 |