Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
Chromosome Location
Chr.1: 16985958 - 17011972 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001141973.2 NP_001135445.1 9 - 10 1060 70
NM_001141974.2 NP_001135446.1 9 - 10 1060 70
NM_022089.3 NP_071372.1 9 - 10 1075 70
XM_005245810.1 XP_005245867.1 9 - 10 1072 70
XM_005245811.1 XP_005245868.1 9 - 10 1060 70
XM_005245812.1 XP_005245869.1 9 - 10 1048 70
XM_005245815.1 XP_005245872.1 9 - 10 1075 70
XM_006710512.1 XP_006710575.1 9 - 10 1057 70
XM_006710513.1 XP_006710576.1 9 - 10 1033 70
XM_011541128.1 XP_011539430.1 9 - 10 1075 70
XM_011541129.1 XP_011539431.1 9 - 10 1075 70
XM_017000844.1 XP_016856333.1 9 - 10 1075 70
XM_017000845.1 XP_016856334.1 9 - 10 1057 70
XM_017000846.1 XP_016856335.1 9 - 10 1033 70
XM_017000847.1 XP_016856336.1 9 - 10 1045 70
XM_017000848.1 XP_016856337.1 9 - 10 1075 70
XM_017000849.1 XP_016856338.1 9 - 10 1060 70
XM_017000850.1 XP_016856339.1 9 - 10 1075 70
GenBank mRNA AK290210.1 - 9 - 10 1016 70
AL354615.1 - 9 - 10 875 70
AY461712.1 - 9 - 10 874 70
BC030267.1 - 9 - 10 1003 70
BC034575.1 - 6 - 7 566 70

View Full Product Details