Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP47
Entrez Gene ID
55031
Gene Name
ubiquitin specific peptidase 47
Gene Aliases
TRFP
Chromosome Location
Chr.11: 11841423 - 11959325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282659.1 NP_001269588.1 12 - 13 2276 82
NM_017944.3 NP_060414.3 11 - 12 2072 82
XM_005252997.1 XP_005253054.1 13 - 14 2336 82
XM_011520196.1 XP_011518498.1 5 - 6 1451 82
XM_017017953.1 XP_016873442.1 13 - 14 2392 82
XM_017017954.1 XP_016873443.1 5 - 6 2599 82
GenBank mRNA AK092290.1 - 13 - 14 1692 82
AK127264.1 - 12 - 13 2072 82
AK309240.1 - 12 - 13 1740 82
BG752249.1 - - 103 82
DA490237.1 - 4 - 5 493 82

View Full Product Details