The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282659.1 | NP_001269588.1 | 5 - 6 | 1358 | 135 |
NM_017944.3 | NP_060414.3 | 4 - 5 | 1154 | 135 | |
XM_005252997.1 | XP_005253054.1 | 6 - 7 | 1418 | 135 | |
XM_017017953.1 | XP_016873442.1 | 6 - 7 | 1474 | 135 | |
GenBank mRNA | AK027362.1 | - | 4 - 5 | 559 | 135 |
AK092290.1 | - | 6 - 7 | 775 | 135 | |
AK127264.1 | - | 4 - 5 | 1154 | 135 | |
AK309240.1 | - | 5 - 6 | 822 | 135 |