Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEZ6
Entrez Gene ID
124925
Gene Name
seizure related 6 homolog
Gene Aliases
BSRPC
Chromosome Location
Chr.17: 28954905 - 29006440 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098635.1 NP_001092105.1 6 - 7 1606 57
NM_001290202.1 NP_001277131.1 6 - 7 1442 57
NM_178860.4 NP_849191.3 6 - 7 1606 57
XM_011524315.1 XP_011522617.1 6 - 7 1606 57
XM_011524317.2 XP_011522619.1 6 - 7 1606 57
GenBank mRNA AF502129.1 - 6 - 7 1694 57
AF502130.1 - 6 - 7 1694 57
AK091522.1 - 6 - 7 1442 57
AK096473.1 - 5 - 6 910 57
AK125377.1 - 6 - 7 1568 57
AK223620.1 - 6 - 7 1568 57
AL834405.1 - 5 - 6 1351 57
AY038048.1 - 6 - 7 1597 57

View Full Product Details