The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029835.2 | NP_001025006.1 | 3 - 4 | 423 | 96 |
NM_001167934.1 | NP_001161406.1 | 2 - 3 | 261 | 96 | |
NM_001167935.1 | NP_001161407.1 | 3 - 4 | 435 | 96 | |
NM_031443.3 | NP_113631.1 | 3 - 4 | 435 | 96 | |
NR_030770.1 | 3 - 4 | 371 | 96 | ||
XM_006715785.3 | XP_006715848.1 | 2 - 3 | 1313 | 96 | |
XM_006715786.3 | XP_006715849.1 | 3 - 4 | 1031 | 96 | |
XM_011515561.2 | XP_011513863.1 | 3 - 4 | 385 | 96 | |
XM_011515562.1 | XP_011513864.1 | 3 - 4 | 416 | 96 | |
XM_011515563.2 | XP_011513865.1 | 2 - 3 | 1313 | 96 | |
XM_011515564.2 | XP_011513866.1 | 2 - 3 | 248 | 96 | |
XM_017012671.1 | XP_016868160.1 | 3 - 4 | 385 | 96 | |
XM_017012672.1 | XP_016868161.1 | 2 - 3 | 1313 | 96 | |
XM_017012673.1 | XP_016868162.1 | 2 - 3 | 216 | 96 | |
GenBank mRNA | AF370392.1 | - | 3 - 4 | 435 | 96 |
AK025807.1 | - | 2 - 3 | 150 | 96 | |
AK098005.1 | - | 3 - 4 | 1031 | 96 | |
AK303216.1 | - | 2 - 3 | 211 | 96 | |
BC004903.2 | - | 3 - 4 | 370 | 96 | |
BC008859.2 | - | 3 - 4 | 370 | 96 | |
BC016832.2 | - | 3 - 4 | 364 | 96 | |
DA420491.1 | - | 3 - 4 | 359 | 96 |