Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS2
Entrez Gene ID
8470
Gene Name
sorbin and SH3 domain containing 2
Gene Aliases
ARGBP2, PRO0618
Chromosome Location
Chr.4: 185585444 - 185956716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145670.1 NP_001139142.1 11 - 12 1710 77
NM_001145674.1 NP_001139146.1 4 - 5 729 77
NM_001145675.1 NP_001139147.1 5 - 6 774 77
NM_001270771.1 NP_001257700.1 13 - 14 1764 77
NM_021069.4 NP_066547.1 10 - 11 1544 77
XM_005263313.1 XP_005263370.1 11 - 12 1714 77
XM_017008748.1 XP_016864237.1 14 - 15 2875 77
XM_017008753.1 XP_016864242.1 14 - 15 2875 77
XM_017008754.1 XP_016864243.1 13 - 14 2830 77
XM_017008755.1 XP_016864244.1 11 - 12 1840 77
XM_017008756.1 XP_016864245.1 10 - 11 1543 77
XM_017008759.1 XP_016864248.1 11 - 12 2436 77
XM_017008762.1 XP_016864251.1 12 - 13 1759 77
XM_017008763.1 XP_016864252.1 11 - 12 1709 77
XM_017008764.1 XP_016864253.1 12 - 13 1690 77
XM_017008765.1 XP_016864254.1 13 - 14 1993 77
XM_017008766.1 XP_016864255.1 11 - 12 1666 77
XM_017008767.1 XP_016864256.1 12 - 13 1969 77
XM_017008768.1 XP_016864257.1 11 - 12 1887 77
XM_017008769.1 XP_016864258.1 11 - 12 1645 77
XM_017008770.1 XP_016864259.1 10 - 11 1621 77
XM_017008772.1 XP_016864261.1 11 - 12 1597 77
XM_017008773.1 XP_016864262.1 12 - 13 1900 77
GenBank mRNA AB018320.1 - 10 - 11 1196 77
AK225327.1 - 11 - 12 1676 77
AK295742.1 - 11 - 12 1115 77
AK296461.1 - 4 - 5 506 77
AK304653.1 - 5 - 6 510 77

View Full Product Details