Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS2
Entrez Gene ID
8470
Gene Name
sorbin and SH3 domain containing 2
Gene Aliases
ARGBP2, PRO0618
Chromosome Location
Chr.4: 185585444 - 185956716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145670.1 NP_001139142.1 18 - 19 2527 153
NM_001145671.2 NP_001139143.1 20 - 21 2316 153
NM_001145672.1 NP_001139144.1 18 - 19 1954 153
NM_001145673.1 NP_001139145.1 20 - 21 2840 153
NM_001145674.1 NP_001139146.1 12 - 13 3127 153
NM_001145675.1 NP_001139147.1 12 - 13 1591 153
NM_001270771.1 NP_001257700.1 21 - 22 4162 153
NM_003603.6 NP_003594.3 20 - 21 2555 153
NM_021069.4 NP_066547.1 18 - 19 3942 153
XM_005263307.1 XP_005263364.1 22 - 23 3434 153
XM_005263308.1 XP_005263365.1 21 - 22 2792 153
XM_005263310.1 XP_005263367.1 21 - 22 2648 153
XM_005263311.1 XP_005263368.1 20 - 21 2633 153
XM_005263312.1 XP_005263369.1 20 - 21 2630 153
XM_005263313.1 XP_005263370.1 18 - 19 2531 153
XM_006714388.1 XP_006714451.1 21 - 22 2723 153
XM_006714390.1 XP_006714453.1 19 - 20 2540 153
XM_017008750.1 XP_016864239.1 24 - 25 3992 153
XM_017008751.1 XP_016864240.1 23 - 24 3908 153
XM_017008752.1 XP_016864241.1 23 - 24 3833 153
XM_017008753.1 XP_016864242.1 21 - 22 3692 153
XM_017008754.1 XP_016864243.1 20 - 21 3647 153
XM_017008758.1 XP_016864247.1 21 - 22 2826 153
XM_017008759.1 XP_016864248.1 18 - 19 3253 153
XM_017008760.1 XP_016864249.1 20 - 21 2699 153
XM_017008762.1 XP_016864251.1 19 - 20 2576 153
XM_017008763.1 XP_016864252.1 18 - 19 2526 153
XM_017008764.1 XP_016864253.1 19 - 20 2507 153
XM_017008765.1 XP_016864254.1 20 - 21 2810 153
XM_017008766.1 XP_016864255.1 18 - 19 2483 153
XM_017008767.1 XP_016864256.1 19 - 20 2786 153
XM_017008768.1 XP_016864257.1 18 - 19 2704 153
XM_017008769.1 XP_016864258.1 18 - 19 2462 153
XM_017008770.1 XP_016864259.1 17 - 18 2438 153
XM_017008771.1 XP_016864260.1 19 - 20 2580 153
XM_017008772.1 XP_016864261.1 18 - 19 2414 153
XM_017008773.1 XP_016864262.1 19 - 20 2717 153
XM_017008774.1 XP_016864263.1 14 - 15 2207 153
XM_017008775.1 XP_016864264.1 23 - 24 3947 153
GenBank mRNA AB018320.1 - 18 - 19 3594 153
AF049884.1 - 20 - 21 2519 153
AK056628.1 - 21 - 22 3780 153
AK225327.1 - 18 - 19 2493 153
AK225812.1 - 20 - 21 2840 153
AK293400.1 - 18 - 19 1954 153
AK296461.1 - 12 - 13 2904 153
AK304653.1 - 12 - 13 1327 153
BC011883.1 - 20 - 21 2283 153

View Full Product Details