Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS2
Entrez Gene ID
8470
Gene Name
sorbin and SH3 domain containing 2
Gene Aliases
ARGBP2, PRO0618
Chromosome Location
Chr.4: 185585444 - 185956716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145670.1 NP_001139142.1 6 - 7 1026 76
NM_001145671.2 NP_001139143.1 5 - 6 554 76
NM_001145672.1 NP_001139144.1 4 - 5 351 76
NM_001270771.1 NP_001257700.1 7 - 8 1035 76
NM_003603.6 NP_003594.3 5 - 6 868 76
NM_021069.4 NP_066547.1 4 - 5 815 76
XM_005263308.1 XP_005263365.1 6 - 7 1030 76
XM_005263310.1 XP_005263367.1 6 - 7 961 76
XM_005263311.1 XP_005263368.1 6 - 7 1030 76
XM_005263312.1 XP_005263369.1 5 - 6 868 76
XM_005263313.1 XP_005263370.1 6 - 7 1030 76
XM_006714388.1 XP_006714451.1 6 - 7 961 76
XM_006714390.1 XP_006714453.1 5 - 6 937 76
XM_017008755.1 XP_016864244.1 5 - 6 1111 76
XM_017008756.1 XP_016864245.1 4 - 5 814 76
XM_017008758.1 XP_016864247.1 5 - 6 980 76
XM_017008760.1 XP_016864249.1 5 - 6 937 76
XM_017008762.1 XP_016864251.1 6 - 7 1030 76
XM_017008763.1 XP_016864252.1 5 - 6 980 76
XM_017008764.1 XP_016864253.1 6 - 7 961 76
XM_017008765.1 XP_016864254.1 7 - 8 1264 76
XM_017008766.1 XP_016864255.1 5 - 6 937 76
XM_017008767.1 XP_016864256.1 6 - 7 1240 76
XM_017008768.1 XP_016864257.1 5 - 6 1158 76
XM_017008769.1 XP_016864258.1 6 - 7 961 76
XM_017008770.1 XP_016864259.1 5 - 6 937 76
XM_017008771.1 XP_016864260.1 4 - 5 818 76
XM_017008772.1 XP_016864261.1 5 - 6 868 76
XM_017008773.1 XP_016864262.1 6 - 7 1171 76
GenBank mRNA AB018320.1 - 4 - 5 467 76
AF049884.1 - 5 - 6 832 76
AF049885.1 - 4 - 5 197 76
AK056628.1 - 7 - 8 653 76
AK225327.1 - 6 - 7 992 76
AK293400.1 - 4 - 5 351 76
AK295742.1 - 5 - 6 386 76
BC011883.1 - 5 - 6 521 76

View Full Product Details