Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS2
Entrez Gene ID
8470
Gene Name
sorbin and SH3 domain containing 2
Gene Aliases
ARGBP2, PRO0618
Chromosome Location
Chr.4: 185585444 - 185956716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145671.2 NP_001139143.1 13 - 14 1492 90
NM_001145672.1 NP_001139144.1 11 - 12 1130 90
NM_001145673.1 NP_001139145.1 13 - 14 2016 90
NM_003603.6 NP_003594.3 13 - 14 1731 90
XM_005263307.1 XP_005263364.1 15 - 16 2610 90
XM_005263308.1 XP_005263365.1 14 - 15 1968 90
XM_005263310.1 XP_005263367.1 14 - 15 1824 90
XM_005263311.1 XP_005263368.1 13 - 14 1809 90
XM_005263312.1 XP_005263369.1 13 - 14 1806 90
XM_006714388.1 XP_006714451.1 14 - 15 1899 90
XM_006714390.1 XP_006714453.1 12 - 13 1716 90
XM_017008738.1 XP_016864227.1 17 - 18 3168 90
XM_017008739.1 XP_016864228.1 18 - 19 3471 90
XM_017008740.1 XP_016864229.1 17 - 18 3164 90
XM_017008741.1 XP_016864230.1 17 - 18 3168 90
XM_017008742.1 XP_016864231.1 16 - 17 3118 90
XM_017008743.1 XP_016864232.1 17 - 18 3099 90
XM_017008744.1 XP_016864233.1 17 - 18 3378 90
XM_017008745.1 XP_016864234.1 16 - 17 3318 90
XM_017008746.1 XP_016864235.1 16 - 17 3009 90
XM_017008747.1 XP_016864236.1 17 - 18 3309 90
XM_017008749.1 XP_016864238.1 17 - 18 2730 90
XM_017008750.1 XP_016864239.1 17 - 18 3168 90
XM_017008751.1 XP_016864240.1 16 - 17 3084 90
XM_017008752.1 XP_016864241.1 16 - 17 3009 90
XM_017008757.1 XP_016864246.1 6 - 7 938 90
XM_017008758.1 XP_016864247.1 14 - 15 2002 90
XM_017008760.1 XP_016864249.1 13 - 14 1875 90
XM_017008771.1 XP_016864260.1 12 - 13 1756 90
XM_017008774.1 XP_016864263.1 7 - 8 1383 90
XM_017008775.1 XP_016864264.1 16 - 17 3123 90
GenBank mRNA AF049884.1 - 13 - 14 1695 90
AF049885.1 - 12 - 13 1135 90
AK056758.1 - 6 - 7 938 90
AK225812.1 - 13 - 14 2016 90
AK293400.1 - 11 - 12 1130 90
BC011883.1 - 13 - 14 1459 90

View Full Product Details