Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS2
Entrez Gene ID
8470
Gene Name
sorbin and SH3 domain containing 2
Gene Aliases
ARGBP2, PRO0618
Chromosome Location
Chr.4: 185585444 - 185956716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145670.1 NP_001139142.1 2 - 3 731 104
NM_001270771.1 NP_001257700.1 3 - 4 736 104
NM_003603.6 NP_003594.3 2 - 3 731 104
XM_005263307.1 XP_005263364.1 2 - 3 731 104
XM_005263308.1 XP_005263365.1 2 - 3 731 104
XM_005263310.1 XP_005263367.1 2 - 3 731 104
XM_005263311.1 XP_005263368.1 2 - 3 731 104
XM_005263312.1 XP_005263369.1 2 - 3 731 104
XM_005263313.1 XP_005263370.1 2 - 3 731 104
XM_006714388.1 XP_006714451.1 2 - 3 731 104
XM_006714390.1 XP_006714453.1 2 - 3 731 104
XM_017008738.1 XP_016864227.1 2 - 3 731 104
XM_017008741.1 XP_016864230.1 2 - 3 731 104
XM_017008743.1 XP_016864232.1 2 - 3 731 104
XM_017008746.1 XP_016864235.1 2 - 3 731 104
XM_017008748.1 XP_016864237.1 2 - 3 731 104
XM_017008749.1 XP_016864238.1 2 - 3 731 104
XM_017008750.1 XP_016864239.1 2 - 3 731 104
XM_017008751.1 XP_016864240.1 2 - 3 731 104
XM_017008752.1 XP_016864241.1 2 - 3 731 104
XM_017008753.1 XP_016864242.1 2 - 3 731 104
XM_017008754.1 XP_016864243.1 2 - 3 731 104
XM_017008760.1 XP_016864249.1 2 - 3 731 104
XM_017008761.1 XP_016864250.1 2 - 3 731 104
XM_017008762.1 XP_016864251.1 2 - 3 731 104
XM_017008764.1 XP_016864253.1 2 - 3 731 104
XM_017008766.1 XP_016864255.1 2 - 3 731 104
XM_017008769.1 XP_016864258.1 2 - 3 731 104
XM_017008770.1 XP_016864259.1 2 - 3 731 104
XM_017008772.1 XP_016864261.1 2 - 3 731 104
XM_017008775.1 XP_016864264.1 2 - 3 731 104
GenBank mRNA AF049884.1 - 2 - 3 695 104
AF049885.1 - 1 - 2 60 104
AK056628.1 - 3 - 4 354 104
AK225327.1 - 2 - 3 697 104

View Full Product Details