Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VNN3
Entrez Gene ID
55350
Gene Name
vanin 3
Gene Aliases
HSA238982
Chromosome Location
Chr.6: 132722787 - 132734765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB435053.1 - 2 - 3 343 71
AK314708.1 - 2 - 3 416 71

View Full Product Details